Learn More
WYK Sorbents LLC SORBENT 2 1/2LB SHAKER 12/CS
SDP
Catalog No. 50465287
Encompass_Preferred
Anti-Slip Absorbent; Inert Mineral that is non-biodegradable that will not leach out (degrade) into the landfill; certified free of Respirable Silica. Effective on heavy viscous liquids
Catalog No. 50-465-287 Supplier WYK Sorbents LLC Supplier No. 500
Anti-Slip Absorbent; Inert Mineral that is non-biodegradable that will not leach out (degrade) into the landfill; certified free of Respirable Silica. Effective on heavy viscous liquids