Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbent, Bulk Silica C8, 10 grams, Solstice

Catalog No. 6002988 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
Packaging:
Fiberboard Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
600-29-88 1 Ea. Fiberboard Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 600-29-88 Supplier Solstice Supplier No. 9940
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bulk Silica C8, 10 grams

  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71889-02-6
Molecular Formula C18H37Cl3O2Si2
Molecular Weight (g/mol) 448.012
InChI Key VYTRPUAMVCWPLO-UHFFFAOYSA-N
PubChem CID 172932
IUPAC Name dioxosilane;trichloro(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl.O=[Si]=O

Specifications

Quantity 1 Ea.
Packaging Fiberboard Box
Grade Analytical
Chemical Name or Material Sorbent, Bulk Silica C8, 10 grams
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.