Learn More
SKC Inc SORBENT TUBE/SILICA GEL 20/PK
SDP
Catalog No. NC9539403
Encompass
TUBE-SORBENT,20PK,6 X 110 mm size,SKC Sorbent Tube, Silica Gel (2,4-Dinitrophenylhydrazine),2-section, 150/300 mg sorbent DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC9539403 Supplier SKC Inc Supplier No. 226119
TUBE-SORBENT,20PK,6 X 110 mm size,SKC Sorbent Tube, Silica Gel (2,4-Dinitrophenylhydrazine),2-section, 150/300 mg sorbent DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”