Learn More
Apexbio Technology LLC SORBIC-ACID-100MG
SDP
Catalog No. 5000568589
Encompass_Preferred
Sorbic acid (CAS No 110-44-1) is a widely researched compound in the biomedical field due to its antimicrobial properties Originating as a natural preservative sorbic acid inhibits the growth of molds yeasts and fungi by interfering with microbial enzymatic functions and weakening cellular membranes Its mechanism involves disrupting the metabolic pathways necessary for microbial energy production In vitro studies demonstrate its inhibitory activity typically ranges from nanomolar to low micromolar concentrations Sorbic acid is commonly applied in various research settings including cell models and animal studies to evaluate its effectiveness and potential therapeutic uses It s frequently utilized in drug screening processes to assess antimicrobial efficacy Experimental concentrations of sorbic acid typically depend on the specific research design allowing flexibility for targeted investigations
Catalog No. 50-005-68589 Supplier Apexbio Technology LLC Supplier No. C6522100
Add to Cart
Add to Cart
Sorbic acid (CAS No 110-44-1) is a widely researched compound in the biomedical field due to its antimicrobial properties Originating as a natural preservative sorbic acid inhibits the growth of molds yeasts and fungi by interfering with microbial enzymatic functions and weakening cellular membranes Its mechanism involves disrupting the metabolic pathways necessary for microbial energy production In vitro studies demonstrate its inhibitory activity typically ranges from nanomolar to low micromolar concentrations Sorbic acid is commonly applied in various research settings including cell models and animal studies to evaluate its effectiveness and potential therapeutic uses It s frequently utilized in drug screening processes to assess antimicrobial efficacy Experimental concentrations of sorbic acid typically depend on the specific research design allowing flexibility for targeted investigations