Learn More
Medchemexpress LLC Sorbic acid | 110-44-1 | 112.13 | 5 G
SDP
Catalog No. 5000388581
Encompass_Preferred
Sorbic acid is an effective, safe, and non-toxic food preservative that can inhibit most molds, yeasts, and certain bacteria. It is orally active and can enhance the growth performance of economically important animals by activating the insulin-like growth factor system (IGF). It can also lead to lipid accumulation by disrupting lipid clearance pathways. This product is for research use only and not sold to patients.
  • Inhibits most molds, yeasts, and certain bacteria.
  • Enhances growth performance in economically important animals.
  • Can lead to lipid accumulation by disrupting lipid clearance pathways.
  • Inhibits growth activity of Aspergillus niger by inducing intracellular acidification.
Catalog No. 50-003-88581 Supplier Medchemexpress LLC Supplier No. HYN06265G
Add to Cart
Add to Cart
Sorbic acid is an effective, safe, and non-toxic food preservative that can inhibit most molds, yeasts, and certain bacteria. It is orally active and can enhance the growth performance of economically important animals by activating the insulin-like growth factor system (IGF). It can also lead to lipid accumulation by disrupting lipid clearance pathways. This product is for research use only and not sold to patients.
  • Inhibits most molds, yeasts, and certain bacteria.
  • Enhances growth performance in economically important animals.
  • Can lead to lipid accumulation by disrupting lipid clearance pathways.
  • Inhibits growth activity of Aspergillus niger by inducing intracellular acidification.