Learn More
Medchemexpress LLC Sorbic acid-13C2 | 114.11 | C4(13C)2H8O2 | 1mg
SDP
Supplier:  Medchemexpress LLC HYW7477511MG
Encompass_Preferred
Sorbic acid-13C2 is the 13C-labeled Sorbic acid (HY-N0626) Sorbic acid is an effective safe and non-toxic food preservative It can effectively inhibit most molds yeasts and certain bacteria Sorbic acid is orally active and can enhance the growth performance of economically important animals (Duroc Landrace Yorkshire piglets) by activating the insulin-like growth factor system (IGF) and it can also lead to lipid accumulation by disrupting lipid clearance pathways[1][2][3][4]
Catalog No. 50-002-70417