Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012455 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2455 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2455 Supplier MilliporeSigma Supelco Supplier No. PHR13671G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 110-44-1
Vapor Pressure 0.01 mmHg (20°C)
Flash Point 127°C
Physical Form Neat
Linear Formula CH3 CH=CHCH=CHCOOH
MDL Number MFCD00002703
Melting Point 132°C to 135°C
Synonym Sorbic acid; alpha-trans-gamma-trans-Sorbic acid; (E,E)-1,3-Pentadiene-1-carboxylic acid; 2,4-Hexadienoic acid; 2 E,4 E-Hexadienoic acid; trans,trans-2,4-Hexadienoic acid; C6:2 n-2,4; NSC 35405; NSC 49103; NSC 50268
Type Certified Reference Material
Quantity 1 g
Formula Weight 112.13
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.