Learn More
ERA WATERS CORP Turbidity, WasteWatR™
SDP
Catalog No. NC1047953
Encompass
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with nephelometric methods
Catalog No. NC1047953 Supplier ERA WATERS CORP Supplier No. 777
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with nephelometric methods