Learn More
Medchemexpress LLC Sorbitan monododecanoate | 1338-39-2 | MFCD00005365 | 99.0% | 346.5 g/mol | C18H34O6 | 1 KG
SDP
Catalog No. 5000283272
Encompass_Preferred
Sorbitan monododecanoate is a sorbitan ester nonionic surfactant used as an emulsifier and pharmaceutical excipient. It acts as a formulation aid to improve solubility, stability, and processing of oil-in-water and water-in-oil systems, and is typically supplied as a viscous, colorless to light-yellow liquid with high purity suitable for excipient use.
  • Nonionic surfactant and emulsifier
  • Improves solubility and stability of formulations
  • Suitable for oil-in-water and water-in-oil emulsions
  • Viscous liquid, colorless to light yellow appearance
  • High purity (99.0%) for excipient applications
  • Compatible with a wide range of formulation ingredients
Catalog No. 50-002-83272 Supplier Medchemexpress LLC Supplier No. HYW1299881KG
Sorbitan monododecanoate is a sorbitan ester nonionic surfactant used as an emulsifier and pharmaceutical excipient. It acts as a formulation aid to improve solubility, stability, and processing of oil-in-water and water-in-oil systems, and is typically supplied as a viscous, colorless to light-yellow liquid with high purity suitable for excipient use.
  • Nonionic surfactant and emulsifier
  • Improves solubility and stability of formulations
  • Suitable for oil-in-water and water-in-oil emulsions
  • Viscous liquid, colorless to light yellow appearance
  • High purity (99.0%) for excipient applications
  • Compatible with a wide range of formulation ingredients