Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sorbitan monododecanoate | 1338-39-2 | MFCD00005365 | 99.0% | 346.5 g/mol | C18H34O6 | 2.5 KG
SDP

Catalog No. 5000283274
Encompass_Preferred

Sorbitan monododecanoate is a sorbitan ester nonionic surfactant and pharmaceutical excipient used as an emulsifier and stabilizer in formulations. It appears as a viscous, colorless to light yellow liquid and is used to improve solubility, stability, and processing in drug formulations and in industrial emulsification applications.

  • Nonionic surfactant and emulsifier.
  • Purity: 99.0%.
  • Viscous, colorless to light yellow liquid.
  • Solubility in DMSO: 100 mg/mL (requires ultrasonication).
  • Chemical formula C18H34O6; molar mass 346.5 g/mol.
  • Available in 2.5 kg package.

Catalog No. 50-002-83274 Supplier Medchemexpress LLC Supplier No. HYW1299882.5KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sorbitan monododecanoate is a sorbitan ester nonionic surfactant and pharmaceutical excipient used as an emulsifier and stabilizer in formulations. It appears as a viscous, colorless to light yellow liquid and is used to improve solubility, stability, and processing in drug formulations and in industrial emulsification applications.

  • Nonionic surfactant and emulsifier.
  • Purity: 99.0%.
  • Viscous, colorless to light yellow liquid.
  • Solubility in DMSO: 100 mg/mL (requires ultrasonication).
  • Chemical formula C18H34O6; molar mass 346.5 g/mol.
  • Available in 2.5 kg package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.