Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sorbitan monododecanoate | 1338-39-2 | MFCD00005365 | 99.0% | 346.5 g/mol | C18H34O6 | 500 G
SDP

Catalog No. 5000283275
Encompass_Preferred

Sorbitan monododecanoate is a nonionic sorbitan ester used as an emulsifier and surfactant in pharmaceutical and cosmetic formulations. It serves as an inactive excipient to improve solubility, stability, and processability of dosage forms, and helps produce homogeneous emulsions and consistent performance in final products.

  • Purity 99.0%.
  • CAS number 1338-39-2.
  • Nonionic surfactant and emulsifier.
  • Improves solubility, stability, and processability of formulations.
  • Suitable as a pharmaceutical and cosmetic excipient.
  • Available in 500 g, 1 kg, and 2.5 kg packages.

Catalog No. 50-002-83275 Supplier Medchemexpress LLC Supplier No. HYW129988500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sorbitan monododecanoate is a nonionic sorbitan ester used as an emulsifier and surfactant in pharmaceutical and cosmetic formulations. It serves as an inactive excipient to improve solubility, stability, and processability of dosage forms, and helps produce homogeneous emulsions and consistent performance in final products.

  • Purity 99.0%.
  • CAS number 1338-39-2.
  • Nonionic surfactant and emulsifier.
  • Improves solubility, stability, and processability of formulations.
  • Suitable as a pharmaceutical and cosmetic excipient.
  • Available in 500 g, 1 kg, and 2.5 kg packages.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.