Learn More
Medchemexpress LLC Sorbitan monooctadecanoate | 1338-41-6 | MFCD00005366 | 95.0% | 430.6 g/mol | C24H46O6 | 1 KG
SDP
Catalog No. 5000282812
Encompass_Preferred
Sorbitan monooctadecanoate is a nonionic surfactant and emulsifying excipient used to stabilize oil-water systems and improve dispersion of lipophilic compounds in formulations. It is commonly applied as an emulsifier, solubilizer, wetting agent, and dispersant in laboratory and formulation settings.
  • Acts as a nonionic emulsifier for oil-in-water systems.
  • Improves solubility of lipophilic compounds.
  • Provides wetting and dispersing action for suspensions.
  • Compatible with a wide range of formulation matrices.
  • Available in multiple pack sizes for laboratory use.
Catalog No. 50-002-82812 Supplier Medchemexpress LLC Supplier No. HYW1295931KG
Sorbitan monooctadecanoate is a nonionic surfactant and emulsifying excipient used to stabilize oil-water systems and improve dispersion of lipophilic compounds in formulations. It is commonly applied as an emulsifier, solubilizer, wetting agent, and dispersant in laboratory and formulation settings.
  • Acts as a nonionic emulsifier for oil-in-water systems.
  • Improves solubility of lipophilic compounds.
  • Provides wetting and dispersing action for suspensions.
  • Compatible with a wide range of formulation matrices.
  • Available in multiple pack sizes for laboratory use.