Learn More
Medchemexpress LLC Sorbitan monooctadecanoate | 1338-41-6 | MFCD00005366 | 95.0% | 430.6 g/mol | C24H46O6 | 500 G
SDP
Catalog No. 5000282715
Encompass_Preferred
Sorbitan monooctadecanoate is a hydrophobic, nonionic surfactant used as a pharmaceutical excipient to stabilize, solubilize, and improve processing of formulations. It is a white to off-white solid commonly employed in emulsions, suspensions, and wetting systems.
  • Functions as an emulsifier and nonionic surfactant.
  • Improves solubility and dispersion of hydrophobic ingredients.
  • Acts as a wetting and suspending agent in formulations.
  • Solid, easy-to-handle form suitable for a range of processing methods.
  • Stable under normal ambient storage for typical shelf life.
Catalog No. 50-002-82715 Supplier Medchemexpress LLC Supplier No. HYW129593500G
Sorbitan monooctadecanoate is a hydrophobic, nonionic surfactant used as a pharmaceutical excipient to stabilize, solubilize, and improve processing of formulations. It is a white to off-white solid commonly employed in emulsions, suspensions, and wetting systems.
  • Functions as an emulsifier and nonionic surfactant.
  • Improves solubility and dispersion of hydrophobic ingredients.
  • Acts as a wetting and suspending agent in formulations.
  • Solid, easy-to-handle form suitable for a range of processing methods.
  • Stable under normal ambient storage for typical shelf life.