Learn More
Medchemexpress LLC Sorbitan monopalmitate | 26266-57-9 | MFCD00080946 | 402.6 g/mol | C22H42O6 | 25 G
SDP
Catalog No. 5000283256
Encompass_Preferred
Sorbitan monopalmitate is a nonionic surfactant and emulsifier used as a pharmaceutical excipient for formulation development. It is commonly used to stabilize emulsions, aid niosome particle preparation, and improve solubility and processability in laboratory-scale formulations. Supplied in small pack sizes appropriate for research and formulation work.
  • Nonionic surfactant and emulsifier for formulation use.
  • Supports niosome and emulsion preparation.
  • Improves solubility and stability of formulations.
  • Suitable for laboratory and small-scale formulation work.
  • Available in small pack sizes for research use.
Catalog No. 50-002-83256 Supplier Medchemexpress LLC Supplier No. HYW12951325G
Sorbitan monopalmitate is a nonionic surfactant and emulsifier used as a pharmaceutical excipient for formulation development. It is commonly used to stabilize emulsions, aid niosome particle preparation, and improve solubility and processability in laboratory-scale formulations. Supplied in small pack sizes appropriate for research and formulation work.
  • Nonionic surfactant and emulsifier for formulation use.
  • Supports niosome and emulsion preparation.
  • Improves solubility and stability of formulations.
  • Suitable for laboratory and small-scale formulation work.
  • Available in small pack sizes for research use.