Learn More
Medchemexpress LLC Sorbitan monostearate | 1338-41-6 | MFCD00005366 | 430.6 g/mol | C24H46O6 | 100 G
SDP
Catalog No. 5000282949
Encompass_Preferred
Sorbitan monooctadecanoate (sorbitan monostearate) is a hydrophobic, nonionic surfactant commonly used as a pharmaceutical excipient. It serves as an emulsifier, solubilizer, wetting agent, and dispersing/suspending aid to improve the stability, solubility, and processability of formulations in pharmaceutical and related applications.
  • Hydrophobic nonionic surfactant
  • Acts as an emulsifier and solubilizer
  • Improves wetting and dispersion of solids
  • Supports stability and processability of formulations
  • White to off-white solid appearance
  • Provided in a 100 G packaging option
Catalog No. 50-002-82949 Supplier Medchemexpress LLC Supplier No. HYW129593100G
Sorbitan monooctadecanoate (sorbitan monostearate) is a hydrophobic, nonionic surfactant commonly used as a pharmaceutical excipient. It serves as an emulsifier, solubilizer, wetting agent, and dispersing/suspending aid to improve the stability, solubility, and processability of formulations in pharmaceutical and related applications.
  • Hydrophobic nonionic surfactant
  • Acts as an emulsifier and solubilizer
  • Improves wetting and dispersion of solids
  • Supports stability and processability of formulations
  • White to off-white solid appearance
  • Provided in a 100 G packaging option