Learn More
Medchemexpress LLC Sorbitan sesquioleate | 8007-43-0 | MFCD00151568 | 100.0% | 1175.70 | C66H126O16 | 250mL
SDP
Catalog No. 5000269484
Encompass_Preferred
Sorbitan Sesquioleate is a synthetic surfactant and emulsifier based on sorbitol and modified oleic acid[1]
Catalog No. 50-002-69484 Supplier Medchemexpress LLC Supplier No. HYW763560250ML
Sorbitan Sesquioleate is a synthetic surfactant and emulsifier based on sorbitol and modified oleic acid[1]