Learn More
Medchemexpress LLC Sorbitan trioleate 500g
SDP
Catalog No. 5000206113
Encompass_Preferred
Sorbitan trioleate is an orally active nonionic surfactant with low toxicity (LD 200 mg/kg) Sorbitan trioleate can be used as an excipient such as surfactant emulsifier lubricant wetting agent dispersant thickener defoamer Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]
Catalog No. 50-002-06113 Supplier Medchemexpress LLC Supplier No. HYW115716500G
Sorbitan trioleate is an orally active nonionic surfactant with low toxicity (LD 200 mg/kg) Sorbitan trioleate can be used as an excipient such as surfactant emulsifier lubricant wetting agent dispersant thickener defoamer Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]