Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitan Trioleate, Spectrum™ Chemical
SDP

Catalog No. 18606941 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-941 4 L Amber Glass Bottle
18-606-942 20 L Poly Drum
18-606-943 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-941 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SPA854LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

SPA85, 26266-58-0, C60H108O8

Spectrum™ Chemical Sorbitan Trioleate

Chemical Identifiers

CAS 26266-58-0
Molecular Formula C60H108O8
Molecular Weight (g/mol) 957.52
InChI Key PRXRUNOAOLTIEF-XDTJCZEISA-N
IUPAC Name 2-[(2R,3S,4R)-4-hydroxy-3-[(9Z)-octadec-9-enoyloxy]oxolan-2-yl]-2-[(9Z)-octadec-9-enoyloxy]ethyl (9Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(OC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC[C@@H](O)[C@@H]1OC(=O)CCCCCCC\C=C/CCCCCCCC

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 4 L
Viscosity Approx. 210 cps
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.