Learn More
Medchemexpress LLC Sorbitan tristearate | 26658-19-5 | MFCD00151165 | ≥98.0% | 963.54 g/mol | C60H114O8 | 1 KG
SDP
Catalog No. 5000283286
Encompass_Preferred
Sorbitan tristearate is a non-ionic surfactant used as a pharmaceutical excipient and emulsifier in formulation development. It is commonly used in topical and cosmetic preparations to improve emulsion stability, enhance skin permeability, and support consistent delivery of active ingredients.
  • Non-ionic surfactant and emulsifier for oil-in-water and water-in-oil systems.
  • Helps improve emulsion stability and product texture.
  • Can influence skin permeability and topical delivery.
  • High purity suitable for formulation and laboratory use.
  • Available in bulk packaging for production-scale applications.
Catalog No. 50-002-83286 Supplier Medchemexpress LLC Supplier No. HYW1311011KG
Sorbitan tristearate is a non-ionic surfactant used as a pharmaceutical excipient and emulsifier in formulation development. It is commonly used in topical and cosmetic preparations to improve emulsion stability, enhance skin permeability, and support consistent delivery of active ingredients.
  • Non-ionic surfactant and emulsifier for oil-in-water and water-in-oil systems.
  • Helps improve emulsion stability and product texture.
  • Can influence skin permeability and topical delivery.
  • High purity suitable for formulation and laboratory use.
  • Available in bulk packaging for production-scale applications.