Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitan Tristearate, Spectrum™ Chemical
SDP

Catalog No. 18613979 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
90 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-979 90 kg Poly Pail
18-613-977 2.5 kg Poly Pail
18-613-976 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-613-979 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SPA6590KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26658-19-5

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 90 kg
Saponification Value 176 - 188
Molecular Weight (g/mol) 963.56
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Sorbitan Tristearate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.