Learn More
U.S. Pharmacopeia Sorbitol, 50-70-4, MFCD00004708, 125 mg
Catalog No. NC9107546
Encompass
Empirical Formula (Hill Notation): C6H14O6, Molecular Weight: 182.17, MP: 98-100 °C, Synonym: D-Glucitol, D-Sorbitol.
Catalog No. NC9107546 Supplier U.S. Pharmacopeia Supplier No. 1617000
Empirical Formula (Hill Notation): C6H14O6, Molecular Weight: 182.17, MP: 98-100 °C, Synonym: D-Glucitol, D-Sorbitol.