Learn More
Apexbio Technology LLC Sorbitol 50-70-4 500mg
SDP
Catalog No. 5000093227
Encompass_Preferred
Sorbitol is a six-carbon sugar alcohol used to modulate cellular osmotic pressure and stabilize protein structures It acts as an osmotic regulator influencing cell-volume control and osmo-responsive signaling pathways In experimental models elevated sorbitol concentrations induce hyperosmotic stress facilitating studies of cellular adaptation apoptosis signaling and molecular stress-response cascades Additionally sorbitol serves as a stabilizing excipient in protein formulations protecting against protein denaturation during freezing thawing or drying Based on these properties sorbitol holds research potential in cellular stress studies and as a stabilizer in biopharmaceutical formulations
Catalog No. 50-000-93227 Supplier Apexbio Technology LLC Supplier No. B2029500
Sorbitol is a six-carbon sugar alcohol used to modulate cellular osmotic pressure and stabilize protein structures It acts as an osmotic regulator influencing cell-volume control and osmo-responsive signaling pathways In experimental models elevated sorbitol concentrations induce hyperosmotic stress facilitating studies of cellular adaptation apoptosis signaling and molecular stress-response cascades Additionally sorbitol serves as a stabilizing excipient in protein formulations protecting against protein denaturation during freezing thawing or drying Based on these properties sorbitol holds research potential in cellular stress studies and as a stabilizer in biopharmaceutical formulations