Learn More
Sigma Aldrich Fine Chemicals Biosciences Sorbitol Dehydrogenase Ass
Catalog No. 502904381
Encompass
Sorbitol Dehydrogenase (SDH) is an enzyme that catalyzes the interconversion of sorbitol and fructose. Elevated blood serum SDH levels indicate liver damage thus SDH plays an important role in the diagnosis of liver disease especially in combination with aminotransferases. SDH levels are also measured to evaluate diabetic complications such as proliferative diabetic retinopathy.
Catalog No. 50-290-4381 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3171KT
Sorbitol Dehydrogenase (SDH) is an enzyme that catalyzes the interconversion of sorbitol and fructose. Elevated blood serum SDH levels indicate liver damage thus SDH plays an important role in the diagnosis of liver disease especially in combination with aminotransferases. SDH levels are also measured to evaluate diabetic complications such as proliferative diabetic retinopathy.