Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sorbitol dehydrogenase-IN-1 | 300551-49-9 | 98.7% | 343.43 | C17H25N7O | 1 MG
SDP

Catalog No. 5000229032
Encompass_Preferred

Potent, orally active sorbitol dehydrogenase inhibitor supplied as a 1 mg solid for research use. It exhibits low-nanomolar inhibitory activity against rat and human sorbitol dehydrogenase and is provided with high stated purity.

  • Potent inhibitory activity: IC50 = 4 nM (rat), 5 nM (human).
  • High purity: 98.7% as stated by the manufacturer.
  • Chemical identity: formula C17H25N7O; molecular weight 343.43.
  • Available as a 1 MG solid and as a 10 mM solution in DMSO for reconstitution.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C for extended storage.

Catalog No. 50-002-29032 Supplier Medchemexpress LLC Supplier No. HY1189801MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, orally active sorbitol dehydrogenase inhibitor supplied as a 1 mg solid for research use. It exhibits low-nanomolar inhibitory activity against rat and human sorbitol dehydrogenase and is provided with high stated purity.

  • Potent inhibitory activity: IC50 = 4 nM (rat), 5 nM (human).
  • High purity: 98.7% as stated by the manufacturer.
  • Chemical identity: formula C17H25N7O; molecular weight 343.43.
  • Available as a 1 MG solid and as a 10 mM solution in DMSO for reconstitution.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.