Learn More
Medchemexpress LLC Sorbitol dehydrogenase-IN-1 | 300551-49-9 | 98.7% | 343.43 | C17H25N7O | 100 MG
SDP
Catalog No. 5000227400
Encompass_Preferred
Sorbitol dehydrogenase-IN-1 is a potent, orally active inhibitor of sorbitol dehydrogenase (SDH) used as a research tool to study SDH-dependent metabolic pathways. It displays low-nanomolar inhibitory activity against both rat and human SDH and is supplied at high purity for biochemical and cellular experiments.
  • Potent sorbitol dehydrogenase inhibition (IC50: rat 4 nM; human 5 nM).
  • High purity (98.7%).
  • Molecular weight 343.43 g/mol.
  • Available in multiple package sizes, including 100 mg.
  • Intended for in vitro and in vivo research use only.
Catalog No. 50-002-27400 Supplier Medchemexpress LLC Supplier No. HY118980100MG
Add to Cart
Add to Cart
Request a Quote
Sorbitol dehydrogenase-IN-1 is a potent, orally active inhibitor of sorbitol dehydrogenase (SDH) used as a research tool to study SDH-dependent metabolic pathways. It displays low-nanomolar inhibitory activity against both rat and human SDH and is supplied at high purity for biochemical and cellular experiments.
  • Potent sorbitol dehydrogenase inhibition (IC50: rat 4 nM; human 5 nM).
  • High purity (98.7%).
  • Molecular weight 343.43 g/mol.
  • Available in multiple package sizes, including 100 mg.
  • Intended for in vitro and in vivo research use only.