Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sorbitol dehydrogenase-IN-1 | 300551-49-9 | 98.7% | 343.43 g·mol⁻¹ | C17H25N7O | 1 ML
SDP

Catalog No. 5000227539
Encompass_Preferred

Sorbitol dehydrogenase-IN-1 is a small-molecule inhibitor of sorbitol dehydrogenase provided as a 10 mM solution in DMSO for laboratory research use. The product is intended for biochemical assays and preclinical studies where enzyme inhibition is required.

  • Provided as a 10 mM solution in DMSO for immediate use.
  • Packaged as a 1 mL vial suitable for small-volume assays.
  • High purity (98.7%) for consistent experimental results.
  • Chemical formula C17H25N7O and molecular weight 343.43 g·mol⁻¹.
  • Solubility and storage recommendations available for in vitro and in vivo formulations.

Catalog No. 50-002-27539 Supplier Medchemexpress LLC Supplier No. HY11898010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sorbitol dehydrogenase-IN-1 is a small-molecule inhibitor of sorbitol dehydrogenase provided as a 10 mM solution in DMSO for laboratory research use. The product is intended for biochemical assays and preclinical studies where enzyme inhibition is required.

  • Provided as a 10 mM solution in DMSO for immediate use.
  • Packaged as a 1 mL vial suitable for small-volume assays.
  • High purity (98.7%) for consistent experimental results.
  • Chemical formula C17H25N7O and molecular weight 343.43 g·mol⁻¹.
  • Solubility and storage recommendations available for in vitro and in vivo formulations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.