Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sorbitol dehydrogenase-IN-1 | 300551-49-9 | 98.7% | 343.43 g/mol | C17H25N7O | 5 MG
SDP

Catalog No. 5000228612
Encompass_Preferred

Sorbitol dehydrogenase-IN-1 is a potent, orally active inhibitor of sorbitol dehydrogenase (SDH) used for biochemical and pharmacological research into SDH-related metabolic pathways. It exhibits low-nanomolar activity against rat and human SDH and is supplied as a high-purity solid and as a ready-to-use solution for laboratory studies.

  • Potent SDH inhibition with low-nanomolar IC50 values.
  • High purity (98.7%) suitable for research assays.
  • Available as a solid powder and as a 10 mM solution in DMSO.
  • Soluble in DMSO (≈40 mg/mL with warming and sonication).
  • Storage-stable when stored under recommended conditions (powder frozen or refrigerated).
  • Intended for in vitro biochemical assays and inhibitor profiling.

Catalog No. 50-002-28612 Supplier Medchemexpress LLC Supplier No. HY1189805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sorbitol dehydrogenase-IN-1 is a potent, orally active inhibitor of sorbitol dehydrogenase (SDH) used for biochemical and pharmacological research into SDH-related metabolic pathways. It exhibits low-nanomolar activity against rat and human SDH and is supplied as a high-purity solid and as a ready-to-use solution for laboratory studies.

  • Potent SDH inhibition with low-nanomolar IC50 values.
  • High purity (98.7%) suitable for research assays.
  • Available as a solid powder and as a 10 mM solution in DMSO.
  • Soluble in DMSO (≈40 mg/mL with warming and sonication).
  • Storage-stable when stored under recommended conditions (powder frozen or refrigerated).
  • Intended for in vitro biochemical assays and inhibitor profiling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.