Learn More
Medchemexpress LLC Sorbitol dehydrogenase-IN-1 | 300551-49-9 | MFCD05260911 | 98.7% | 343.43 | C17H25N7O | 25 MG
SDP
Catalog No. 5000229033
Encompass_Preferred
Sorbitol dehydrogenase-IN-1 is a potent, orally active sorbitol dehydrogenase inhibitor used for biochemical and pharmacological research, including studies of the polyol pathway and diabetes-related complications. It is supplied as a research small-molecule powder with multiple pack sizes for flexible dosing.
  • Low-nanomolar potency (IC50 ≈ 4-5 nM) against human and rat sorbitol dehydrogenase.
  • Suitable for in vitro and in vivo studies of sorbitol dehydrogenase activity and related metabolic pathways.
  • High purity for research applications (manufacturer lists purity ~98.7%).
  • Well characterized: CAS 300551-49-9, formula C17H25N7O, molecular weight ~343.43 g·mol-1.
Catalog No. 50-002-29033 Supplier Medchemexpress LLC Supplier No. HY11898025MG
Sorbitol dehydrogenase-IN-1 is a potent, orally active sorbitol dehydrogenase inhibitor used for biochemical and pharmacological research, including studies of the polyol pathway and diabetes-related complications. It is supplied as a research small-molecule powder with multiple pack sizes for flexible dosing.
  • Low-nanomolar potency (IC50 ≈ 4-5 nM) against human and rat sorbitol dehydrogenase.
  • Suitable for in vitro and in vivo studies of sorbitol dehydrogenase activity and related metabolic pathways.
  • High purity for research applications (manufacturer lists purity ~98.7%).
  • Well characterized: CAS 300551-49-9, formula C17H25N7O, molecular weight ~343.43 g·mol-1.