Learn More
Medchemexpress LLC Sorbitol dehydrogenase-IN-1 | 300551-49-9 | MFCD05260911 | 98.7% | 343.43 g/mol | C17H25N7O | 10 MG
SDP
Catalog No. 5000227402
Encompass_Preferred
Sorbitol dehydrogenase-IN-1 is a potent, orally active sorbitol dehydrogenase inhibitor used in preclinical research into diabetes and cardiovascular disease. It is supplied as a white to off-white solid with high purity suitable for biochemical and cellular assays.
  • Potent sorbitol dehydrogenase inhibitor with low-nanomolar activity (rat and human IC50 ~4-5 nM).
  • High purity (98.7%) appropriate for biochemical assays.
  • White to off-white solid, stable under recommended storage conditions.
  • Available in small research sizes, for example, 10 MG.
  • Molecular weight approximately 343.43 g/mol and formula C17H25N7O.
Catalog No. 50-002-27402 Supplier Medchemexpress LLC Supplier No. HY11898010MG
Sorbitol dehydrogenase-IN-1 is a potent, orally active sorbitol dehydrogenase inhibitor used in preclinical research into diabetes and cardiovascular disease. It is supplied as a white to off-white solid with high purity suitable for biochemical and cellular assays.
  • Potent sorbitol dehydrogenase inhibitor with low-nanomolar activity (rat and human IC50 ~4-5 nM).
  • High purity (98.7%) appropriate for biochemical assays.
  • White to off-white solid, stable under recommended storage conditions.
  • Available in small research sizes, for example, 10 MG.
  • Molecular weight approximately 343.43 g/mol and formula C17H25N7O.