Learn More
NEOGEN Sorbitol MacConkey Agar, 500 g
SDP
Catalog No. 502015636
Encompass_Preferred
Sorbitol MacConkey Agar is used for the isolation of pathogenic Escherichia coli in a laboratory setting. Sorbitol MacConkey Agar is not intended for use in the diagnosis of disease or other conditions in humans. Sorbitol MacConkey Agar is based on the formula by Rappaport and Henig. Originally developed for isolating enteropathogenic (EPEC) serotypes, O11 and O55, this medium is recommended for the isolation and differentiation of enterohemorrhagic E. coli O157:H7. Serotype O157 has been implicated in serious foodborne diseases. MacConkey Agar W/ Sorbitol contains sorbitol instead of lactose for differentiating enteropathogenic E. coli serotypes; these strains are typically sorbitol negative. MacConkey Agar W/ Sorbitol is recommended for food testing.For more information visit https://www.neogen.com/solutions/Microbiology/sorbitol-macconkey-agar
Catalog No. 50-201-5636 Supplier NEOGEN Supplier No. 700003460
Sorbitol MacConkey Agar is used for the isolation of pathogenic Escherichia coli in a laboratory setting. Sorbitol MacConkey Agar is not intended for use in the diagnosis of disease or other conditions in humans. Sorbitol MacConkey Agar is based on the formula by Rappaport and Henig. Originally developed for isolating enteropathogenic (EPEC) serotypes, O11 and O55, this medium is recommended for the isolation and differentiation of enterohemorrhagic E. coli O157:H7. Serotype O157 has been implicated in serious foodborne diseases. MacConkey Agar W/ Sorbitol contains sorbitol instead of lactose for differentiating enteropathogenic E. coli serotypes; these strains are typically sorbitol negative. MacConkey Agar W/ Sorbitol is recommended for food testing.For more information visit https://www.neogen.com/solutions/Microbiology/sorbitol-macconkey-agar