Learn More
Electron Microscopy Sciences AMS Series Sorbothane Balance Platform, 40 x 50 cm, Stainless Steel Surface
SDP
Catalog No. 501930080
Encompass_Preferred
The AMS tables incorporate Sorbothane isolators, giving excellent isolation with a natural frequency of less than 7Hz. Our isolators considerably outperform both simple air cushion and rubber isolators, shortening stabilisation times and speeding up measurements.
Size: 40 x 50 cm
Catalog No. 50-193-0080 Supplier Electron Microscopy Sciences Supplier No. 687040
The AMS tables incorporate Sorbothane isolators, giving excellent isolation with a natural frequency of less than 7Hz. Our isolators considerably outperform both simple air cushion and rubber isolators, shortening stabilisation times and speeding up measurements.
Size: 40 x 50 cm