Learn More
Sorbent Technologies Inc Sorbtech Rocket - TLC Development Chamber
SDP
Catalog No. NC1938409
Encompass_Preferred
Sorbtech Rocket TLC Chamber for 2.5 x 7.5cm TLC Plates - an innovative, TLC small-scale development chamber that delivers more efficient, effective, and safer handling of TLC. Solvent consumption is reduced by 80-85%. Rf overruns are eliminated using a reduced solvent. Overall operational time is faster. Results are far more consistent test to test. The TLC plate is always kept in the same orientation. Rocket shape and white background provide easy visualization. Can be used with glass, plastic, and aluminum backed plates.
Catalog No. NC1938409 Supplier Sorbent Technologies Inc Supplier No. TLCROCKET
Sorbtech Rocket TLC Chamber for 2.5 x 7.5cm TLC Plates - an innovative, TLC small-scale development chamber that delivers more efficient, effective, and safer handling of TLC. Solvent consumption is reduced by 80-85%. Rf overruns are eliminated using a reduced solvent. Overall operational time is faster. Results are far more consistent test to test. The TLC plate is always kept in the same orientation. Rocket shape and white background provide easy visualization. Can be used with glass, plastic, and aluminum backed plates.