Learn More
Cayman Chemical SORC13trIfluoroACTAte sal 10mg
SDP
Catalog No. 502909463
Encompass_Preferred
A peptide TRPV6 antagonist; inhibits calcium flux in HEK293 cells expressing human TRPV6 (EC50 = 14 nM); reduces tumor volume in an SKOV3 ovarian cancer mouse xenograft model
Catalog No. 50-290-9463 Supplier Cayman Chemical Supplier No. 3676410 MG
A peptide TRPV6 antagonist; inhibits calcium flux in HEK293 cells expressing human TRPV6 (EC50 = 14 nM); reduces tumor volume in an SKOV3 ovarian cancer mouse xenograft model