Learn More
Cayman Chemical SOR-C13 (trifluoroacetate salt), Purity: ≥98%, 50 mg
SDP
Catalog No. 502913263
Encompass_Preferred
A peptide TRPV6 antagonist; inhibits calcium flux in HEK293 cells expressing human TRPV6 (EC50 = 14 nM); reduces tumor volume in an SKOV3 ovarian cancer mouse xenograft model
Catalog No. 50-291-3263 Supplier Cayman Chemical Supplier No. 3676450 MG
A peptide TRPV6 antagonist; inhibits calcium flux in HEK293 cells expressing human TRPV6 (EC50 = 14 nM); reduces tumor volume in an SKOV3 ovarian cancer mouse xenograft model