Learn More
Aviva Systems Biology Corp SORD Antibody - N-terminal region (ARP61128_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61128_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SORD. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SORD. Alternate names: SORD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-287-262