Learn More
Medchemexpress LLC Sordarin sodium | 463356-00-5 | 99.0% | 514.58 | 1 MG
SDP
Catalog No. 5000438139
Encompass_Preferred
Sordarin sodium is an eEF2 inhibitor and antibiotic. Its function depends on the diphthamide modification on eEF2. It inhibits protein synthesis by preventing the binding of eEF2 to the ribosome complex and exhibits antifungal activity.
  • eEF2 inhibitor
  • Antibiotic
  • Antifungal activity
  • White to off-white solid appearance
  • Inhibits protein synthesis
  • Active against *Saccharomyces cerevisiae*
Catalog No. 50-004-38139 Supplier Medchemexpress LLC Supplier No. HY1263961MG
Sordarin sodium is an eEF2 inhibitor and antibiotic. Its function depends on the diphthamide modification on eEF2. It inhibits protein synthesis by preventing the binding of eEF2 to the ribosome complex and exhibits antifungal activity.
  • eEF2 inhibitor
  • Antibiotic
  • Antifungal activity
  • White to off-white solid appearance
  • Inhibits protein synthesis
  • Active against *Saccharomyces cerevisiae*