Learn More
Alkali Scientific Sorensen Buffer 500mL Bottle , 6/CS
SDP
Catalog No. 5000662782
Encompass_Preferred
Sorensen Buffer is a classic phosphate buffer system widely used in biological and clinical laboratories to maintain stable pH conditions during staining and analytical procedures. It is particularly important in hematology when preparing blood smears using Wright or Giemsa staining methods. The pH of the buffer influences how cellular components absorb stains, affecting the color and contrast of microscopic preparations. By maintaining a consistent pH, Sorensen Buffer ensures reliable staining results and improved visualization of blood cell structures. The standardized formulation eliminates variations that may occur when using unbuffered water, helping laboratories achieve consistent diagnostic and research results.
Catalog No. 50-006-62782 Supplier Alkali Scientific Supplier No. SSOB00
Sorensen Buffer is a classic phosphate buffer system widely used in biological and clinical laboratories to maintain stable pH conditions during staining and analytical procedures. It is particularly important in hematology when preparing blood smears using Wright or Giemsa staining methods. The pH of the buffer influences how cellular components absorb stains, affecting the color and contrast of microscopic preparations. By maintaining a consistent pH, Sorensen Buffer ensures reliable staining results and improved visualization of blood cell structures. The standardized formulation eliminates variations that may occur when using unbuffered water, helping laboratories achieve consistent diagnostic and research results.