Learn More
Electron Microscopy Sciences A Phosphate buffered mixed salt prepared from sodium phosphate monobasic and sodium phosphate dibasic. Osmolarity is 300 mosmols.
SDP
Supplier:  Electron Microscopy Sciences 11601104L
Encompass_Preferred
Sorensen's Phosphate Buffer is also know as phosphate mixed sodium salts. The osmolarity of a 0.1 M at a pH 7.2 is 226 mosmols; with the addition of 0.18 M of sucrose to the solution, the osmolarity raises to 425 mosmols. Maximum shelf life after receipt is 3 months.
Catalog No. NC2754990