Learn More
Electron Microscopy Sciences Sorensen's Phosphate Buffer, 0.15M, 1 L
SDP
Catalog No. 50980234
Encompass_Preferred
A Phosphate buffered mixed salt prepared from sodium phosphate monobasic and sodium phosphate dibasic. Maximum shelf life after receipt: 3 months. Standard pH 7.2; 0.2M is supplied. Otherwise, specify pH when ordering.
Catalog No. 50-980-234 Supplier Electron Microscopy Sciences Supplier No. 1168210
A Phosphate buffered mixed salt prepared from sodium phosphate monobasic and sodium phosphate dibasic. Maximum shelf life after receipt: 3 months. Standard pH 7.2; 0.2M is supplied. Otherwise, specify pH when ordering.