Learn More
Andwin Scientific SORENSON 10340 OneTouch™ Pipet Tips, 200ul, 4800/case
SDP
Catalog No. NC0717081
Encompass_Preferred
The sterile OneTouch™ Pipet Tips feature accuracy and high-quality performance. The product is compatible with a huge variety of pipettors and is free of RNase, DNase, and pyrogens. The OneTouch tips provide a leak proof seal and require a minimal force to be ejected. Tips have a volume capacity of 200 µL (1–330 µL Range). This product is packaged in plastic racks.
Catalog No. NC0717081 Supplier Andwin Scientific Supplier No. SORENSON 10340
The sterile OneTouch™ Pipet Tips feature accuracy and high-quality performance. The product is compatible with a huge variety of pipettors and is free of RNase, DNase, and pyrogens. The OneTouch tips provide a leak proof seal and require a minimal force to be ejected. Tips have a volume capacity of 200 µL (1–330 µL Range). This product is packaged in plastic racks.