Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Cap Strips

Catalog No. 05408217 Shop All Fisherbrand Products
Change view
Click to view available options
Array:
1 x 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Array
05-408-217 1 x 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-408-217

missing translation for 'validMainframeMsg'

Optically clear

  • 8-Strip
  • Fit PCR tubes

TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Array 1 x 8
Color Clear
For Use With (Equipment) PCR tubes
No. of Wells 8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.