Learn More
Electron Microscopy Sciences Sorensens Phosphate Buffer, 0.2M, 1 L
SDP
Catalog No. 50980229
Encompass_Preferred
A Phosphate buffered mixed salt prepared from sodium phosphate monobasic and sodium phosphate dibasic, pH 7.2.
Catalog No. 50-980-229 Supplier Electron Microscopy Sciences Supplier No. 1160010
A Phosphate buffered mixed salt prepared from sodium phosphate monobasic and sodium phosphate dibasic, pH 7.2.