Learn More
Electron Microscopy Sciences Sorensons Phosphate Buffer, 0.2M Standard pH 7.2 500 ML
SDP
Catalog No. 50980228
Encompass_Preferred
(Phosphate Mixed Sodium Salts)
Sorensen's Phosphate Buffer is also know as phosphate mixed sodium salts. The osmolarity of a 0.1 M at a pH 7.2 is 226 mosmols; with the addition of 0.18 M of sucrose to the solution, the osmolarity raises to 425 mosmols. Maximum shelf life after receipt is 3 months.
Standard pH: 7.2
Catalog No. 50-980-228 Supplier Electron Microscopy Sciences Supplier No. 1160005
(Phosphate Mixed Sodium Salts)
Sorensen's Phosphate Buffer is also know as phosphate mixed sodium salts. The osmolarity of a 0.1 M at a pH 7.2 is 226 mosmols; with the addition of 0.18 M of sucrose to the solution, the osmolarity raises to 425 mosmols. Maximum shelf life after receipt is 3 months.
Standard pH: 7.2