Learn More
Sigma Aldrich Fine Chemicals Biosciences SorensonTM Dolphin microce
Catalog No. 502901666
Encompass
The Dolphin microcentrifuge tubes are Ideal design for pelletizing samples. They have a positive seal design which allows for repeated cap closures and will not pop open under pressure when boiling.RCf rating of 15 000 x gfits all standard rotors (for 2.0 mL tubes)Certified RNase/DNase freeCertified PCR Inhibitor freeCertified Human DNA freeCertified Pyrogen free
Catalog No. 50-290-1666 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7175331000EA
The Dolphin microcentrifuge tubes are Ideal design for pelletizing samples. They have a positive seal design which allows for repeated cap closures and will not pop open under pressure when boiling.RCf rating of 15 000 x gfits all standard rotors (for 2.0 mL tubes)Certified RNase/DNase freeCertified PCR Inhibitor freeCertified Human DNA freeCertified Pyrogen free