Learn More
Sigma Aldrich Fine Chemicals Biosciences SorensonTM low binding sta
Catalog No. 502905896
Encompass
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Catalog No. 50-290-5896 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7196174800EA
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.