Learn More
Cayman Chemical Isoorientin, Purity: ≥95%, CAS: 4261-42-1, 50 mg
SDP
Catalog No. 502934890
Encompass_Preferred
A flavonoid with diverse biological activities; induces apoptosis in and reduces cell viability of PATU-8988 and PANC-1 pancreatic cancer cells; reduces expression of COX-2, IL-6, 5-LO, and TNF-α in RAW 264.7 cells; reduces carrageenan-induced paw edema and air pouch blood vessel swelling in mice at 20 mg/kg; increases stomach levels of SOD and GSH and reduces ulcer area in a rat model of indomethacin-induced gastric ulcers
Catalog No. 50-293-4890 Supplier Cayman Chemical Supplier No. 2686250 MG
A flavonoid with diverse biological activities; induces apoptosis in and reduces cell viability of PATU-8988 and PANC-1 pancreatic cancer cells; reduces expression of COX-2, IL-6, 5-LO, and TNF-α in RAW 264.7 cells; reduces carrageenan-induced paw edema and air pouch blood vessel swelling in mice at 20 mg/kg; increases stomach levels of SOD and GSH and reduces ulcer area in a rat model of indomethacin-induced gastric ulcers