Learn More
Medchemexpress LLC SORT-PGRN interactio 100mg | 100957-85-5 | 258.32 g/mol | C15H18N2O2 | 100 MG
SDP
Catalog No. 5000226475
Encompass_Preferred
SORT-PGRN interaction inhibitor 1 is a small-molecule research reagent that inhibits the sortilin-progranulin protein-protein interaction with an in vitro IC50 of 2 μM. It is supplied as a solid and as DMSO solutions for biochemical and cellular assays.
  • Potent inhibition of the sortilin-progranulin interaction (IC50 = 2 μM).
  • Small molecule with formula C15H18N2O2 and molecular weight 258.32 g/mol.
  • High reported purity (~99.4%) suitable for biochemical studies.
  • Available as solid and preprepared DMSO solutions for assay flexibility.
  • Typically provided in multiple pack sizes to support screening and follow-up experiments.
Catalog No. 50-002-26475 Supplier Medchemexpress LLC Supplier No. HY115213100MG
SORT-PGRN interaction inhibitor 1 is a small-molecule research reagent that inhibits the sortilin-progranulin protein-protein interaction with an in vitro IC50 of 2 μM. It is supplied as a solid and as DMSO solutions for biochemical and cellular assays.
  • Potent inhibition of the sortilin-progranulin interaction (IC50 = 2 μM).
  • Small molecule with formula C15H18N2O2 and molecular weight 258.32 g/mol.
  • High reported purity (~99.4%) suitable for biochemical studies.
  • Available as solid and preprepared DMSO solutions for assay flexibility.
  • Typically provided in multiple pack sizes to support screening and follow-up experiments.