Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SORT-PGRN interactio 250mg | 100957-85-5 | 250 MG
SDP

Catalog No. 5000226919
Encompass_Preferred

SORT-PGRN interaction inhibitor 1 is a small-molecule research compound that disrupts the sortilin-progranulin interaction and is used in biochemical and cell-based assays to study progranulin biology.

  • Inhibits sortilin-progranulin interaction with IC50 2 μM.
  • High purity, 99.37%.
  • Molecular formula C15H18N2O2 and molecular weight 258.32.
  • Soluble in DMSO at 100 mg/mL for in vitro applications.
  • Available in common research quantities including 250 MG.
  • Powder is stable at -20°C for long-term storage; follow SDS and COA for handling and preparation.

Catalog No. 50-002-26919 Supplier Medchemexpress LLC Supplier No. HY115213250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SORT-PGRN interaction inhibitor 1 is a small-molecule research compound that disrupts the sortilin-progranulin interaction and is used in biochemical and cell-based assays to study progranulin biology.

  • Inhibits sortilin-progranulin interaction with IC50 2 μM.
  • High purity, 99.37%.
  • Molecular formula C15H18N2O2 and molecular weight 258.32.
  • Soluble in DMSO at 100 mg/mL for in vitro applications.
  • Available in common research quantities including 250 MG.
  • Powder is stable at -20°C for long-term storage; follow SDS and COA for handling and preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.