Learn More
Aviva Systems Biology Corp SORT1 Antibody - C-terminal region (ARP64630_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64630_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SORT1. It was validated on Western Blot. . Alternate names: Gp95, NT3ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-320