Learn More
BPS Bioscience Inc Sortase A, Octamutant, His-Tag, 50 µg
SDP
Supplier:  BPS Bioscience Inc 72518
Encompass
Staphylococcus aureus sortase A Octamutant (P94R, D124G, D160N, D165A, Y187L, E189R, K190E, K196T), a.a 26-206(end) GenBank Accession No. AF162687 with N-terminal His tag, MW = 22 kDa, expressed in an E. coli expression system.
Catalog No. NC1646520